• USA Home
  • N5913 - Monoclonal Anti-NPDC-1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
N5913 Sigma-Aldrich

Monoclonal Anti-NPDC-1 antibody produced in mouse

~1 mg/mL, clone 16E4, purified immunoglobulin, buffered aqueous glycerol solution

Synonym: Anti-Neuronal proliferation, differentiation and control protein 1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Gene Regulation and Expression, Antibodies to Transcription Regulators,
conjugate   unconjugated
clone   16E4, monoclonal
biological source   mouse
application(s)   western blot: 1:1,000 using lysate of cells overexpressing NPDC-1
species reactivity   human
mol wt   antigen 34-40 kDa
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG1
antibody product type   primary antibodies
concentration   ~1 mg/mL
UniProt accession no.   Q9NQX5
Gene Information   human ... NPDC1(56654)

Description

General description

Neural proliferation, differentiation and control-1 (NPDC-1) is expressed in the lungs and brain. The gene encoding this 34kDa protein is localized on chromosome 9q34.3.

Immunogen

peptide SLGSPVSSDPVHMS.

Physical form

solution in 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, and 3 mM sodium azide.

Biochem/physiol Actions

Neural proliferation, differentiation and control-1 (NPDC-1) interacts with cyclins and other proteins of the cell cycle. It is a regulator of retinoid signaling.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?