• USA Home
  • N6509 - 4-Nitrophenyl β-D-gentiobioside

EMAIL THIS PAGE TO A FRIEND
N6509 Sigma-Aldrich

4-Nitrophenyl β-D-gentiobioside

  • CAS Number 16790-33-3

  • Empirical Formula (Hill Notation) C18H25NO13

  • Molecular Weight 463.39

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?