• USA Home
  • N8158 - Monoclonal Anti-Nicotinic Acetylcholine Receptor (α7 Subunit) antibody produced in rat

EMAIL THIS PAGE TO A FRIEND
N8158 Sigma-Aldrich

Monoclonal Anti-Nicotinic Acetylcholine Receptor (α7 Subunit) antibody produced in rat

~5 mg/mL, clone mAb 319, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

conjugate   unconjugated
clone   mAb 319, monoclonal
biological source   rat
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  western blot: suitable
species reactivity   human, rat, chicken
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG1
concentration   ~5 mg/mL
Gene Information   chicken ... CHRNA7(374001)
human ... CHRNA7(1139)
rat ... Chrna7(25302)

Description

Specificity

The antibody recognizes an epitope with amino acids 365-384 of the α7 subunit. Reacts well with chick, but is less reactive with mammalian.

Immunogen

bacterially expressed α7 subunit of the nicotinic acetylcholine receptors.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in 10 mM phosphate buffered saline, containing 10 mM sodium azide.

Legal Information

Sold with the permission of the Salk Institute

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?