• USA Home
  • N8661 - 5′-Nucleotidase from Crotalus atrox venom

EMAIL THIS PAGE TO A FRIEND
N8661 Sigma-Aldrich

5′-Nucleotidase from Crotalus atrox venom

lyophilized powder, ≥200 units/mg protein

Synonym: 5′-Ribonucleotide phosphohydrolase

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Biochemicals and Reagents, Enzyme Class Index, Enzymes, Inhibitors, and Substrates More...
form   lyophilized powder
specific activity   ≥200 units/mg protein
shipped in   dry ice
storage temp.   −20°C

Description

Unit Definition

One unit will hydrolyze 1.0 μmole of inorganic phosphorus from adenosine 5′-monophosphate per min at pH 9.0 at 37 °C.

Physical form

Lyophilized powder balance primarily trehalose, HEPES and CaCl2

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Related Content

Enzymes & Proteins

Committed to developing and providing breakthrough problem-solving reagents and solutions, explore our comprehensive offering of enzymes and protein reagents and kits with the confidence that you wil...
Keywords: Cell signaling, Diagnostic, Enzyme-linked immunosorbent assay, Immunohistochemistry, Metabolism, Molecular biology, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?