EMAIL THIS PAGE TO A FRIEND
N9413 Sigma-Aldrich

NSC 295642

≥98% (HPLC), solid

Synonym: Bis-[chloro[phenylmethyl [1-(2-pyridinyl)ethylidene]hydrazinecarbodithioato-N2,N2′,S1’]-copper], Cu(II) complex of the Schiff base product of condensation of S-benzyl dithiocarbazate and 2-acetylpyridine, [1-(2-pyridinyl)ethylidene]-Hydrazinecarbodithioic acid, phenylmethyl ester, copper complex

  • CAS Number 77111-29-6

  • Empirical Formula (Hill Notation) C30H28Cl2Cu2N6S4

  • Molecular Weight 798.84

  •  PubChem Substance ID 329818694

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Kinase/Phosphatase Biology,
Quality Level   100
assay   ≥98% (HPLC)
form   solid
color   dark green
solubility   DMSO: >5 mg/mL
SMILES string   C\C(=N/N=C(/SCc1ccccc1)S[Cu]Cl)c2ccccn2
InChI   1S/C15H15N3S2.ClH.Cu/c1-12(14-9-5-6-10-16-14)17-18-15(19)20-11-13-7-3-2-4-8-13;;/h2-10H,11H2,1H3,(H,18,19);1H;/q;;+2/p-2/b17-12+;;
InChI key   MGXCRQNCPJBYKM-CWQUOYFRSA-L

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

NSC 295642 is a potent inhibitor of cell migration. Cell migration is a basic biological process involved in a range of normal and pathological events, including wound healing, embryonic and tissue development, immune function and inflammation, angiogenesis and tumor metastasis. Therefore, cell migration can be adapted as an important biological tool to monitor several physiological processes as well as development of cancer. Accordingly, a potent inhibitor of cell migration, such as NSC 295642 can elucidate the intrinsic molecular mechanism of many biochemical events.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?