• USA Home
  • NEUR1051 - Neuronal Medium - 250mL

EMAIL THIS PAGE TO A FRIEND
NEUR1051 Sigma-Aldrich

Neuronal Medium - 250mL

Purchase

Properties

Related Categories Cell Biology, General Stem Cell Biology, Induced Pluripotent Stem Cells and Reagents, Media for iPSC-derived Differentiated Neural Cells, Stem Cell Biology,
sterility   sterile
form   liquid
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −20°C

Description

Preparation Note

See technical bulletin.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Neural Stem Cell FAQs

The use of neural stem cells (NSCs) in biomedical research is becoming increasingly popular, resulting in breakthrough studies rejected the longstanding belief that neuronal tissue is incapable of re...
Keywords: Cell culture, Central Nervous System, Centrifugation, Clinical, Diffusion, Digestions, Diseases, Gene expression, Immunocytochemistry, Neurodegenerative Diseases, Parkinson Disease

Related Content

Neural Stem Cell Culture

Neural Stem Cell Lines Neural Stem Cell Media and Supplements Neural Stem Cell Markers Stem Cell Culture Protocols
Keywords: Cell biology, Cell culture, Central Nervous System, Cryopreservation, Diseases, Dopamine agents, Environmental, Gene expression, Growth factors, Neuroscience, Phase transitions, Stem cell biology, Transcription

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?