EMAIL THIS PAGE TO A FRIEND
O1755 Supelco

Oxazepam solution

drug standard, 1.0 mg/mL in methanol

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Benzodiazepin Analoga Structure, Benzodiazepines,
grade   drug standard
drug control   Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   −20°C
SMILES string   OC1N=C(c2ccccc2)c3cc(Cl)ccc3NC1=O
InChI   1S/C15H11ClN2O2/c16-10-6-7-12-11(8-10)13(9-4-2-1-3-5-9)18-15(20)14(19)17-12/h1-8,15,20H,(H,17,19)
InChI key   ADIMAYPTOBDMTL-UHFFFAOYSA-N
Gene Information   human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Antianxiety Drugs on Ascentis® Express C18 vs. Standard C18, Demonstrating High Throughput on Ascentis Express

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Antianxiety Drugs on Ascentis® Express C18 vs. Standard C18, Demonstrating High Throughput on Ascentis Express
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Antianxiety Drugs on Ascentis® Phenyl

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Antianxiety Drugs on Ascentis® Phenyl
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Anxiolytic Drugs on Discovery® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Anxiolytic Drugs on Discovery® C18
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Anxiolytic Drugs on SUPELCOSIL™ LC-8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Anxiolytic Drugs on SUPELCOSIL™ LC-8
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Benzodiazepines on Ascentis® Express C18, Acetonitrile Mobile Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Benzodiazepines on Ascentis® Express C18, Acetonitrile Mobile Phase
Keywords: Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Benzodiazepines on Ascentis® Express Phenyl-Hexyl with Acetonitrile Mobile Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Benzodiazepines on Ascentis® Express Phenyl-Hexyl with Acetonitrile Mobile Phase
Keywords: Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Benzodiazepines on Ascentis® Express Phenyl-Hexyl, Methanol Mobile Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Benzodiazepines on Ascentis® Express Phenyl-Hexyl, Methanol Mobile Phase
Keywords: Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Oxazepam Enantiomers on Astec® CHIROBIOTIC® T

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Oxazepam Enantiomers on Astec® CHIROBIOTIC® T
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Oxazepam Enantiomers on Astec® CYCLOBOND® I 2000 DNP

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Oxazepam Enantiomers on Astec® CYCLOBOND® I 2000 DNP
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

LC/MS Analysis of Benzodiazepines on Ascentis® Phenyl (Analyte Mix 1)

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of Benzodiazepines on Ascentis® Phenyl (Analyte Mix 1)
Keywords: Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Mass spectrometry, Pharmaceutical

RP-HPLC Analysis of Benzodiazepines on Ascentis® Express Biphenyl

From our library of Articles, Sigma-Aldrich presents RP-HPLC Analysis of Benzodiazepines on Ascentis® Express Biphenyl
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

UHPLC Analysis of Diazepam and Metabolites on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Diazepam and Metabolites on Ascentis® Express C18
Keywords: Chromatography, Clinical

UHPLC Analysis of Diazepam and Metabolites on Titan™ C18, Competitive Comparison

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Diazepam and Metabolites on Titan™ C18, Competitive Comparison
Keywords: Anti-depressants, Anticonvulsants, Chromatography, Clinical, Mass spectrometry, Metabolites, Pharmaceutical

UHPLC/MS Analysis of Benzodiazepines and Glucuronide Metabolites on Titan™ C18

From our library of Articles, Sigma-Aldrich presents UHPLC/MS Analysis of Benzodiazepines and Glucuronide Metabolites on Titan™ C18
Keywords: High performance liquid chromatography, Mass spectrometry, Metabolites, Separation, Solvents

UHPLC/MS Analysis of Diazepam and Metabolites on Titan™ C18, Comparison of Flow Rate and Column Length

From our library of Articles, Sigma-Aldrich presents UHPLC/MS Analysis of Diazepam and Metabolites on Titan™ C18, Comparison of Flow Rate and Column Length
Keywords: Anti-depressants, Anticonvulsants, Chromatography, Solvents

UHPLC/MS/MS Analysis of Benzodiazepines and Glucuronide Metabolites in Urine on Titan™ C18 after SPE using Supel™-Select HLB, β-Glucuronidase Enzyme Digestion

From our library of Articles, Sigma-Aldrich presents UHPLC/MS/MS Analysis of Benzodiazepines and Glucuronide Metabolites in Urine on Titan™ C18 after SPE using Supel™-Select HLB, β-Glucuronidase Enzyme Digestion
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Mass spectrometry, Metabolites, Pharmaceutical, Sample preparations, Solid phase extractions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?