EMAIL THIS PAGE TO A FRIEND
O2389 Sigma-Aldrich

OMDM-1

≥97% (HPLC), powder

Synonym: (N-[(S)-2-Hydroxy-1-(4-hydroxybenzyl)-ethyl]-octadec-9-enoamide

  • Empirical Formula (Hill Notation) C27H45NO3

  • Molecular Weight 431.65

  •  MDL number MFCD05865240

  •  PubChem Substance ID 329818902

  •  NACRES NA.77

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Neurobiology,
Quality Level   100
assay   ≥97% (HPLC)
form   powder
color   off-white
solubility   DMSO: soluble ~24 mg/mL
storage temp.   room temp
SMILES string   CCCCCCCC\C=C/CCCCCCCC(=O)N[C@H](CO)Cc1ccc(O)cc1
InChI   1S/C27H45NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-27(31)28-25(23-29)22-24-18-20-26(30)21-19-24/h9-10,18-21,25,29-30H,2-8,11-17,22-23H2,1H3,(H,28,31)/b10-9-/t25-/m0/s1
InChI key   ICDMLAQPOAVWNH-JRUKXMRZSA-N

Description

Biochem/physiol Actions

Potent, selective inhibitor of anadamide cellular uptake with low affinity for CB1 and CB2 receptors, no activity at VR1 receptor or fatty acid amide hydrolase (FAAH); stable to enzymatic hydrolysis by rat brain homogenates.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?