• USA Home
  • OGS3100 - PSF-OXB20-NH2-PELB-6HIS-TEV - PELB SECRETION AND 6 HIS TAG PLASMID

EMAIL THIS PAGE TO A FRIEND
OGS3100 Sigma-Aldrich

PSF-OXB20-NH2-PELB-6HIS-TEV - PELB SECRETION AND 6 HIS TAG PLASMID

plasmid vector for molecular cloning

Synonym: cloning vector, expression vector, molecular cloning vector, plasmid vector, plasmid, snapfast vector, vector

  •  NACRES NA.85

Purchase

Properties

Related Categories Cloning and Expression, Molecular Biology, SnapFast Cloning Vectors, SnapFast Vectors for Bacterial Host
form   buffered aqueous solution
mol wt   size 3953 bp
conjugate   6-His tagged
Origin of replication   pUC (500 copies)
Peptide cleavage   TEV
Peptide tag location   N-terminal
Promoter   Promoter name: OXB20
Promoter activity: constitutive
Promoter type: bacterial
Secretion signal   PelB
bacteria selection   kanamycin
reporter gene   none
shipped in   ambient
storage temp.   −20°C

Description

General description

This plasmid is designed to express tagged proteins in E. coli. The plasmid contains a constitutive promoter (OXB20) derived from the region upstream of the E. coli RecA gene. It does not require induction or any additional components for activity. It is the strongest of the bacterial promoters that we provide and this high level of expression can cause expression problems with some proteins with poor solubility. For this reason we sell a range of bacterial promoters with different expression levels (OXB1(low)>OXB20(high)) that can be provided with the peptide tags in this plasmid on request.

About the Peptide Tag:This plasmid contains an Pectate Lyase B (PelB) secretory signal peptide (SP) to allow proteins to be exported from the cytosol. During translocation from the cytosol the signal peptide is removed from the protein by endogenous proteases. This plasmid also contains a secondary Hexa-Histidine (6His) protein tag. The sequence of this tag is: HHHHHHWe provide a range of dual peptide tag plasmids. This is because some peptide tags provide specific biological properties (e. g., small molecule affinity new epitopes solubility or protein secretion) that are not provided by others.

About the Cleavage Tag: This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a TEV cleavage tag. The protein sequence of the cleavage tag is: ENLYFQG. Cleavage occurs between the Glu and Gly residues. TEV is often reported to have better specificity for its recognition site compared to EKT Thrombin or Faxtor Xa.

Promoter Expression Level: This plasmid contains a constitutive bacterial promoter that does not require induction. It is the strongest bacterial promoter we sell and this can cause solubility and expression problems with some proteins. We also offer a range of other bacterial promoters that are compatible with this plasmid and are available on request.

Sequence

Quick-reference Plasmid Map

Please select the file type you require. For reference most cloning programs will import a .gb (Genbank) file and will show all of the plasmids features automatically when downloaded and imported.

Genebank Vector Sequence File

FASTA Vector Sequence File

Full Plasmid Map

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxfordgenetics.com.

Other Notes

Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics. Find out more at Oxford Genetics - Sigma's partner for cloning and expression vectors for molecular biology and synthetic biology applications.

Legal Information

Oxford Genetics is a trademark of Oxford Genetics Ltd

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Genome Compiler

Sigma® Life Science is using our strengths and building on the strengths of others to provide the best tools to synthetic biology researchers. We are happy to participate in Genome Compiler's free, w...
Keywords: Cloning

How to Choose a SnapFast™ Expression Vector

With >1600 vectors to choose from, you have many options when planning your cloning constructs. Finding the perfect expression vector starts with understanding what you need, and what the options are.
Keywords: Cloning, Gene expression, Genetics, Immunoprecipitation, Individual protein Immunoprecipitation, Methylations, Purification, Western blot

Plasmid Product Nomenclature

The SnapFast system is a versatile plasmid cloning platform that provides a range of functional DNA sequences in an easy to clone format. We currently have hundreds of pre-designed DNA sections that ...
Keywords: Catalysis, Cloning, Digestions, PAGE

Primers for Plasmid Sequencing

We have designed a range of forward and reverse sequencing primers that allow you to sequence any insert that you make into a particular position within any of our SnapFast™ expression vectors. Other...
Keywords: Cloning, Gas chromatography, Gene expression, Melting, Sequencing

Restriction Site Positions and Functions

Click on the relevant restriction site to learn more about its function in the SnapFast plasmid system.
Keywords: Antibiotics, Cloning, Diagnostic, Gene expression, PAGE, Peptide synthesis, Phosphorylations, Polymerase chain reaction, Purification, Recombination, Sequencing

SnapFast™ Cloning & Expression Vectors Selection Charts

With over 1600 vectors to choose from, your options may seem endless. For help choosing the right vector for your research needs, consult the tables below.
Keywords: Cloning, Gene expression, Genetics, Molecular biology, Transcription

What is a DNA Component?

Our plasmids are composed of both a core vector backbone and a series of DNA components. Our plasmid platform is all built around the same core backbone, which means that the DNA components within ea...
Keywords: Adsorption, Antibiotics, Apoptosis, Cell culture, Cell disruption, Centrifugation, Chromatography, Cloning, Dialysis, Enzyme activity, Flow cytometry, Gene expression, Genetics, Immobilization, Immunoprecipitation, Nucleic acid denaturation, Precipitation, Purification, Transcription, Transfection, Western blot

Protocols

Cloning the Gene-of-Interest into a Plasmid Vector

Genetic engineering is used in thousands of laboratories around the world. Given its importance it is remarkable that cloning strategies for many of the popular DNA components are not standardised. C...
Keywords: Antibiotics, Buffers, Cloning, Degradations, Dialysis, Digestions, Gene expression, Genetic, Genetics, Individual protein Immunoprecipitation, Molecular biology, Nucleic acid annealing, Peptide synthesis, Phosphorylations, Polymerase chain reaction, Precipitation, Purification, Sequencing, Substitutions, transformation

Quick Start Plasmid Vector Guide

SnapFast™ Vectors are supplied in solution as 5µg plasmid DNA in TE buffer, at ambient temperature. If you are not ready to use your DNA right away, store this in the -20 °C for long-term storage, or...
Keywords: Cloning, Genetics, PAGE, Purification, Transfection, transformation

Restriction Enzyme Cloning Manual

Molecular Cloning is a set of techniques that are used by molecular biologists to insert a gene-of-interest into a vector capable of replication within the target cell. Once the gene is being express...
Keywords: AGE, Cloning, Digestions, Electrophoresis, Gel electrophoresis, Gene expression, Genetic, Genetics, Molecular biology, Molecular biology techniques, Nucleic acid annealing, Peptide synthesis, Phase transitions, Phosphorylations, Polymerase chain reaction, Precipitation, transformation

Selecting Correctly Expressing Recombinants

Blue / White colony screening is a strategy to quickly and easily distinguish between recombinant and non-recombinant colonies. It requires a special vector and a special strain of E. coli. It is par...
Keywords: AGE, Amplification, Antibiotics, Cell disruption, Centrifugation, Cloning, Degradations, Detergents, Diagnostic, Digestions, Electrophoresis, Gel electrophoresis, Molecular biology, Peptide synthesis, Phase transitions, Polymerase chain reaction, Precipitation, Purification, Sequencing, Transfection, transformation

SnapFast™ Cloning & Expression Vectors FAQs

Where do I find the sequences and maps of a SnapFast™ plasmid and insert? How can I confirm the orientation of my insert if I use only one insertion site? How have you removed unwanted restriction si...
Keywords: Buffers, Cloning, Diagnostic, Digestions, Gene expression, Genetics, PAGE, Peptide synthesis, Polymerase chain reaction, Sequencing, transformation

Transforming E. coli with Engineered Plasmid

Inoue and colleagues developed this method in 1990. It works well for many strains commonly used in cloning. The original method calls for growing the overnight E. coli cultures at 18°C. We find that...
Keywords: Antibiotics, Centrifugation, Cloning, Peptide synthesis, Phase transitions, transformation

Related Content

Synthetic Biology Resources

We are using our stregnths to help build the field of Synthetic Biology. Below is a collection of links to our favorite resources. In addtion to these resources, we also offer thousands of products f...
Keywords: Cell culture, Cloning, Gene expression, Genetic, Genetics, Metabolic Pathways, Metabolites, Molecular biology, Molecular biology techniques, Photosynthesis, Poisons, Polymerase chain reaction - quantitative, Transcription, Transfection

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

OGS10 PSF-CMV-KAN - KANAMYCIN RESISTANCE PLASMID, plasmid vector for molecular cloning
OGS103 PSF-CMV-RLUC - CMV RENILLA LUCIFERASE PLASMID, plasmid vector for molecular cloning
OGS1120 PSF-CMV-PURO-COOH-TEV-10HIS - C-TERMINAL 10HIS TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS158 PSF-CMV-UB-BETAGAL ASCI - BETA GALACTOSIDASE PLASMID, plasmid vector for molecular cloning
OGS1913 PSF-TEF1-COOH-TEV-10HIS - C-TERMINAL 10HIS TAG YEAST PLASMID, plasmid vector for molecular cloning
OGS207 PSF-OXB20-BETAGAL - BACTERIAL BETA GALACTOSIDASE PLASMID, plasmid vector for molecular cloning
OGS2767 PSF-OXB20-COOH-TEV-10HIS - C-TERMINAL 10HIS TAG BACTERIAL PLASMID, plasmid vector for molecular cloning
OGS2828 PSF-OXB20-NH2-FLAG®-6HIS-EKT - N-TERMINAL FLAG® AND 6 HIS DUAL TAG BACTERIAL PLASMID, plasmid vector for molecular cloning
OGS394 PSF-CMV-PGK-PURO - DUAL PROMOTER PUROMYCIN PLASMID, plasmid vector for molecular cloning
OGS412 PSF-OXB20-FLUC - BACTERIAL LUCIFERASE PLASMID, plasmid vector for molecular cloning
OGS502 PSF-LACI - LACI PROMOTER BACTERIAL VECTOR, plasmid vector for molecular cloning
OGS505 PSF-CAG-KAN - CAG PROMOTER VECTOR, plasmid vector for molecular cloning
OGS553 PSF-OXB1 - WEAK STRENGTH BACTERIAL PROMOTER PLASMID, plasmid vector for molecular cloning
OGS588 PSF-CMV-BLAST - CMV PROMOTER BLASTICIDIN RESISTANT PLASMID, plasmid vector for molecular cloning
PP2356 Mammalian Promoter Vector Set, plasmid vectors for molecular cloning
PP2393 Mammalian FLAG® Tag Vector Set, plasmid vectors for molecular cloning
CMC0001 SIG10 Chemically Competent Cells, for protein expression and DNA plasmid production
CMC0003 SIG10 HIGH Electrocompetent Cells, for protein expression and DNA plasmid production
CMC0007 SIG10 5a Chemically Competent Cells, for DNA plasmid production
CMC0014 BL21(DE3) Chemically Competent Cells, for protein expression
CMC0017 OverExpress C41(DE3) Chemically Competent Cells, for the highest protein expression

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?