• USA Home
  • P0374 - Anti-PAT antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
P0374 Sigma-Aldrich

Anti-PAT antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-Phosphinotricin Acetyl Transferase, Bar gene

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, P-PA, Plant Biology, Primary Antibodies More...
biological source   rabbit
antibody form   affinity isolated antibody
antibody product type   primary antibodies
clone   polyclonal
form   buffered aqueous solution
mol wt   antigen mol wt 21 kDa
species reactivity   plant
application(s)   western blot: 2-4 μg/mL using extract (cytosolic fraction) of leaves from transgenic Nicotiana tabacum expressing the PAT gene for Streptomyces hygroscopicus
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Immunogen

synthetic peptide corresponding to amino acids 141-157 located near the C-terminus of Streptomyces hygroscopicus PAT, conjugated to KLH.

Specificity

The antibody recognizes bacterial PAT expressed in transgenic tobacco plants.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary and Secondary Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay needs. Use the Antibody...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

P0249 Anti-PAT (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?