• USA Home
  • P1052 - Protein A–Acrylic beads from Staphylococcus aureus

EMAIL THIS PAGE TO A FRIEND
P1052 Sigma

Protein A–Acrylic beads from Staphylococcus aureus

lyophilized powder, 250 μm

Purchase

Properties

form   lyophilized powder
matrix   acrylic beads
matrix activation   oxirane
matrix attachment   free amino groups
matrix spacer   3 atoms
particle size   250 μm
capacity   ~10 mg/mL binding capacity (human IgG)
storage temp.   −20°C

Description

Quantity

Swelling: 1 g swells to approx. 6 ml

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?