EMAIL THIS PAGE TO A FRIEND
P3894 Sigma-Aldrich

Brij® S2

Synonym: Polyethylene glycol octadecyl ether, Polyoxyethylene (2) stearyl ether

Purchase

Properties

description   non-ionic
mp   44-45 °C (lit.)
density   0.893 g/mL at 25 °C (lit.)
InChI   1S/C20H41O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h2-20H2,1H3
InChI key   PSEGVHKUPXNGGJ-UHFFFAOYSA-N

Description

Legal Information

Brij is a registered trademark of Croda International PLC

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
WI6300000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?