• USA Home
  • P4129 - Potassium borohydride

EMAIL THIS PAGE TO A FRIEND
P4129 Sigma

Potassium borohydride

Synonym: Potassium tetrahydroborate

Purchase

Properties

Related Categories Borohydrides, Chemical Synthesis, Reduction, Synthetic Reagents
InChI Key   ICRGAIPBTSPUEX-UHFFFAOYSA-N
mp   500 °C (dec.) (lit.)
density   1.18 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

RIDADR 
UN 1870 4.3/PG 1
WGK Germany 
-
RTECS 
TS7525000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

455571 Potassium borohydride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?