• USA Home
  • P4801 - L-Proline-UL-14C hydrochloride

EMAIL THIS PAGE TO A FRIEND
P4801 Sigma

L-Proline-UL-14C hydrochloride

≥98% (Radiochemical Purity, HPLC), aqueous ethanol solution

Synonym: (S)-(−)-Proline-UL-14C

  • CAS Number 4305-67-3

  • Empirical Formula (Hill Notation) C5H9NO2

  • Molecular Weight 115.13

  •  MDL number MFCD00069672

  •  PubChem Substance ID 24898500

Purchase

Properties

InChI Key   ONIBWKKTOPOVIA-RHRFEJLCSA-N
assay   ≥98% (Radiochemical Purity, HPLC)
form   aqueous ethanol solution
extent of labeling   200-300 mCi per mmol
storage temp.   2-8°C

Description

Packaging

Combi-vial

Physical form

Aqueous solution containing 2% ethanol

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?