• USA Home
  • P4816 - Potassium thiocyanate

EMAIL THIS PAGE TO A FRIEND
P4816 Sigma-Aldrich

Potassium thiocyanate

suitable for photographic applications

Synonym: Potassium rhodanide

  • CAS Number 333-20-0

  • Linear Formula KSCN

  • Molecular Weight 97.18

  •  Beilstein/REAXYS Number 3594799

  •  EC Number 206-370-1

  •  MDL number MFCD00011413

Purchase

Properties

InChI Key   ZNNZYHKDIALBAK-UHFFFAOYSA-M
mp   173 °C (lit.)
suitability   suitable for photographic applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
XL1925000

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?