• USA Home
  • P6141 - Protease from Bacillus polymyxa

EMAIL THIS PAGE TO A FRIEND
P6141 Sigma

Protease from Bacillus polymyxa

Type IX, ~1.0 unit/mg solid

Purchase

Description

General description

Enzyme Commision (E.C.) 3.4.24.x

Other Notes

Metalloenzyme

Unit Definition

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent), unless otherwise indicated.

Protocols & Applications

Non-specific Protease Activity Assay Video

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Protocols

Enzymatic Assay of Protease Using Casein As a Substrate

To standardize a procedure for the enzymatic assay of Protease using Casein as a substrate at Sigma-Aldrich St. Louis.

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?