EMAIL THIS PAGE TO A FRIEND
P7359 Sigma-Aldrich
Primer Premier
Purchase
Properties
Related Categories Computer Software, Labware, Life Science software More...
Description
General description
A comprehensive primer design tool
• PCR primer design- design primer pairs for PCR and multiplex experiments.
• Cross species- Amplify sequences from multiple species.
• Pathogenic detection- Locate oligos in highly conserved regions.
• Allele specific- Exclusively amplify a single member of a set of related sequences.
• Degenerate- Start with an amino acid sequence, reverse translate and design oligos in regions of low degeneracy automatically.
• Nested/multiplex- Checking homology among multiple candidate oligos.
• Restriction enzyme and motif analysis- Select from the database of over 800 enzymes and 200 common motifs.
• Long PCR- Handles sequences up to 50 KB.
System Requirements
Windows 98 / 2000 / ME / XP
Legal Information
Primer Premier is a trademark of Premier Biosoft International
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?