EMAIL THIS PAGE TO A FRIEND
P7391 Sigma-Aldrich

Potassium nitrite

~97% (titration)

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Inorganic Salts, Potassium, Potassium Salts,
assay   ~97% (titration)
mp   350 °C (dec.) (lit.)
SMILES string   [K+].[O-]N=O
InChI   1S/K.HNO2/c;2-1-3/h;(H,2,3)/q+1;/p-1
InChI key   BXNHTSHTPBPRFX-UHFFFAOYSA-M
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1488 5.1 / PGII
WGK Germany 
3
RTECS 
TT3750000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?