• USA Home
  • P7682 - Potassium bicarbonate

EMAIL THIS PAGE TO A FRIEND
P7682 Sigma-Aldrich

Potassium bicarbonate

suitable for insect cell culture

Synonym: Potassium hydrogen carbonate

Purchase

Properties

Related Categories Cell Culture, Chemical Synthesis, Companion Products and Reagents, Inorganic Salts, Insect Platform,
application(s)   cell culture | insect: suitable
SMILES string   [K+].OC([O-])=O
InChI   1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1
InChI key   TYJJADVDDVDEDZ-UHFFFAOYSA-M

Description

Application

Use in insect cell culture applications to control pH.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?