• USA Home
  • P7864 - Sartorius Proline® mechanical adjustable volume pipettor
EMAIL THIS PAGE TO A FRIEND
P7864 Sigma
Sartorius Proline® mechanical adjustable volume pipettor
20-200 μL volume
Purchase
Properties
Related Categories BioHit Pipettors, Labware, Liquid Handling Equipment, Manufacturer Browser, Pipettes, Pipettors and Tips,
feature   electronic: no
  single-channel
mfr. no.   Sartorius 720070
measuring range   20-200 μL volume
volume   200 μL
Description
General description
Manually-operated air displacement digital pipettors dispense liquids accurately and safety in educational, medical research, and industrial laboratories. The hand grip and smooth plunger action of the pipettors ensures comfortable handling. Pipettors feature an ergonomically designed support, tip ejector, and lightweight construction. The volume-setting mechanism provides secure setting of the desired volume. Large, clear digital display set volume. Innovative construction and robust housing result in trouble-free operation and maintenance. 3 years warranty.
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?