• USA Home
  • P7897 - Peruvoside from Thevetia neriifolia

EMAIL THIS PAGE TO A FRIEND
P7897 Sigma-Aldrich

Peruvoside from Thevetia neriifolia

~90%, powder

Synonym: (3β,5β)-3-(6-Deoxy-3-O-methyl-α-L-glucopyranosyl]oxyl)-14-hydroxy-19-oxocard-20(22)-enolide

Purchase

Properties

InChI Key   PMTSPAGBAFCORP-UHFFFAOYSA-N
assay   ~90%
form   powder
color   light yellow
storage temp.   2-8°C

Description

Biochem/physiol Actions

Cardiac glycoside with the same inotropic and chronotropic effects on heart function as ouabain.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?