EMAIL THIS PAGE TO A FRIEND
P8041 Sigma-Aldrich

Potassium chloride

suitable for plant cell culture, ≥99%

Purchase

Properties

assay   ≥99%
application(s)   cell culture | plant: suitable
mp   770 °C (lit.)
density   1.98 g/mL at 25 °C (lit.)
SMILES string   [Cl-].[K+]
InChI   1S/ClH.K/h1H;/q;+1/p-1
InChI key   WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Description

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
RTECS 
TS8050000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?