EMAIL THIS PAGE TO A FRIEND
P8269 Sigma

Potassium iodate

≥99.0%

Purchase

Properties

InChI Key   JLKDVMWYMMLWTI-UHFFFAOYSA-M
assay   ≥99.0%
mp   560 °C (lit.)
density   3.93 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1479 5.1 / PGII
WGK Germany 
3
RTECS 
NN1350000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?