• USA Home
  • P8550 - Pentaethylene glycol monododecyl ether

EMAIL THIS PAGE TO A FRIEND
P8550 Sigma

Pentaethylene glycol monododecyl ether

≥98% (GC)

Synonym: C12E5, Dodecyl pentaethylene glycol ether, Dodecylpentaglycol, Polyoxyethylene (5) lauryl ether

  • CAS Number 3055-95-6

  • Linear Formula CH3(CH2)11(OCH2CH2)5OH

  • Molecular Weight 406.60

  •  Beilstein Registry Number 1797921

  •  EC Number 221-281-8

  •  MDL number MFCD00043064

Purchase

Properties

description   non-ionic
InChI Key   LAPRIVJANDLWOK-UHFFFAOYSA-N
assay   ≥98% (GC)
density   0.963 g/mL at 20 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
SB0333332

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

76437 Pentaethylene glycol monododecyl ether, BioXtra, ≥98.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?