• USA Home
  • P8675 - Hexaethylene glycol monododecyl ether

EMAIL THIS PAGE TO A FRIEND
P8675 Sigma-Aldrich

Hexaethylene glycol monododecyl ether

semisolid

Synonym: C12E6, Dodecylhexaglycol, Polyoxyethylene (6) lauryl ether

  • CAS Number 3055-96-7

  • Linear Formula CH3(CH2)11(OCH2CH2)6OH

  • Molecular Weight 450.65

  •  Beilstein Registry Number 1715367

  •  EC Number 221-282-3

  •  MDL number MFCD00042660

  •  PubChem Substance ID 329820332

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Culture, Detergents, Detergents A to M, Detergents by Applications,
description   non-ionic
InChI Key   OJCFEGKCRWEVSN-UHFFFAOYSA-N
form   semisolid
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
MP4780500

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

52044 Hexaethylene glycol monododecyl ether, BioXtra, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?