• USA Home
  • PLA0289 - Rabbit anti-AS160 Antibody, Affinity Purified

EMAIL THIS PAGE TO A FRIEND
PLA0289 Sigma-Aldrich

Rabbit anti-AS160 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym: AS160, Akt substrate of 160 kDa, DKFZp779C0666, KIAA0603, NIDDM5, TBC (Tre-2; BUB2; CDC16) domain-containing protein, TBC1 domain family, member 4

  •  NACRES NA.41

Purchase

Properties

Related Categories Antibodies, Antibodies for Proximity Ligation Assays, Individual Antibodies More...
biological source   rabbit
application(s)   immunoprecipitation (IP): 2- 10 μg/mg
  western blot: 1:2,000-1:10,000
species reactivity   human
shipped in   wet ice
storage temp.   2-8°C
antibody form   affinity purified immunoglobulin
Quality Level   100
antibody product type   primary antibodies
grade   Powered by Bethyl Laboratories, Inc.
accession no.   NP_055647.2
Gene Information   rabbit ... AS160(9882)

Description

Immunogen

The epitope recognized by PLA0289 maps to a region between residue 1248 and 1298 of human Akt Substrate of 160 kDa using the numbering given in entry NP_055647.2 (GeneID 9882).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Other Notes

The Akt substrate of 160 kDa (AS160) is a substrate of Akt kinase activity and is a GTPase activating protein (GAP) for Rab GTPases. AS160 is encoded by the TBC1D4 (TBC1 domain family, member 4) gene.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?