• USA Home
  • PRS1123 - Anti-Eotaxin antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
PRS1123 Sigma-Aldrich

Anti-Eotaxin antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-Chemokine (C-C motif) ligand 11

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Chemokines, Antibodies to Cytokines and Growth Factors,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunocytochemistry: suitable
  immunofluorescence: suitable
  indirect ELISA: suitable
  western blot: suitable
species reactivity   human
mol wt   predicted mol wt 9-18 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   IgG fraction of antiserum
Quality Level   100
antibody product type   primary antibodies
UniProt accession no.   P51671
Gene Information   human ... CCL11(6356)

Description

Immunogen

a peptide corresponding to amino acids near the carboxy-terminus of human eotaxin.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Linkage

The action of this antibody can be blocked using blocking peptide SBP1123.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?