• USA Home
  • PRS2449 - Anti-P53AIP1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
PRS2449 Sigma-Aldrich

Anti-P53AIP1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-Tumor protein p53 regulated apoptosis inducing protein 1, Anti-p53-regulated apoptosis-inducing protein 1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies with Corresponding Blocking Peptides, N-P, P-PA,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunocytochemistry: suitable
  indirect ELISA: suitable
  western blot: suitable
species reactivity   human
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
UniProt accession no.   Q9HCN2
Gene Information   human ... TP53AIP1(63970)

Description

Immunogen

a 15 amino acid peptide from near the amino-terminus of human p53AIP1.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Linkage

The action of this antibody can be blocked using blocking peptide SBP2449.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?