• USA Home
  • PRS4827 - Anti-Integrin α4 (ab2) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
PRS4827 Sigma-Aldrich

Anti-Integrin α4 (ab2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-α4 subunit of VLA-4 receptor, Anti-CD49d, Anti-IA4, Anti-ITGA4, Anti-Integrin, α4 (antigen CD49D, α4 subunit of VLA-4 receptor)

Purchase

Properties

Related Categories Alphabetical Index, Analytical Cytology, Antibodies, Antibodies for Cell Biology, Antibodies for Immune Cell Signaling and Blood,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunofluorescence: suitable
  indirect ELISA: suitable
  western blot: suitable
species reactivity   rat, human, mouse
mol wt   predicted mol wt 17 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
UniProt accession no.   P13612
Gene Information   human ... ITGA4(3676)

Description

Immunogen

a 16 amino acid peptide from near the center of human Integrin alpha 4.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Linkage

The action of this antibody can be blocked using blocking peptide SBP4827.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?