EMAIL THIS PAGE TO A FRIEND
PZ0356 Sigma-Aldrich

PF-06291874

≥98% (HPLC)

Synonym: N-[4-[(1S)-1-[3,5-Dimethyl-4-[4-(trifluoromethyl)-1H-pyrazol-1-yl]phenoxy]butyl]benzoyl]--alanine

  • CAS Number 1393124-08-7

  • Empirical Formula (Hill Notation) C26H28F3N3O4

  • Molecular Weight 503.51

  •  MDL number MFCD30343861

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones,
Quality Level   100
assay   ≥98% (HPLC)
form   powder
mfr. no.   Pfizer®
color   white to beige
solubility   DMSO: 10 mg/mL, clear
originator   Pfizer
storage temp.   room temp

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

PF-06291874 is an orally active, potent and selective glucagon receptor antagonist. PF-06291874 exhibits a robust glucose reductions in subjects with type 2 diabetes mellitus.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Pfizer is a registered trademark of Pfizer, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?