• USA Home
  • QPREST28734 - SILuPrEST KANK2

EMAIL THIS PAGE TO A FRIEND
QPREST28734 Sigma-Aldrich

SILuPrEST KANK2

SILuPrESTs Powered by Atlas Antibodies, buffered aqueous solution

Synonym: Ankyrin repeat domain-containing protein 25, Matrix-remodeling-associated protein 3, SRC-1-interacting protein

Purchase

Properties

Related Categories Mass Spectrometry, Molecular Biology, Proteomics, SILuPrEST K - L, SILuPrEST Mass Spectrometry Standards More...
recombinant   expressed in E. coli LysA ArgA BL21(DE3)
assay   >80% (SDS-PAGE)
form   buffered aqueous solution
mol wt   predicted mol wt 31kDa including tags
purified by   immobilized metal affinity chromatography (IMAC)
packaging   pkg of 1nmol × 5 vials
storage condition   avoid repeated freeze/thaw cycles
concentration   (Lot specific concentration determined by ESI-MS/MS using highly pure and amino acid analyzed His6ABP as internal standard for targeted quantification of the labeled His6ABP-containing SILuPrEST will be stated on the Certificate of Analysis)
immunogen sequence   SQHIPTAEGASGSNTEEEIRMELSPDLISACLALEKYLDNPNALTERELKVAYTTVLQEWLRLACRSDAHPELVRRHLVTFRAMSARLLDYVVNIADSNGNTALHYSVSHANFPVVQ
UniProt accession no.   Q63ZY3
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... KANK2(25959)

Description

General description

Lys, Arg 13C and 15N metabolically labeled recombinant human protein fragment

Application

Internal standard in MS-based quantitative proteomics

Physical form

Heavy proteins are provided in solution of 1M Urea PBS, pH 7.4

Preparation Note

The protein solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.

Analysis Note

Isotopic Label Incorporation

The SILuPrEST standards are produced using metabolic labeling with heavy isotope labeled (15N, 13C) Lysine and Arginine residues, with greater than 99% isotopic label incorporation.

A purification and quantification tag (QTag) consisting of a hexahistidine (His6) sequence followed by an Albumin Binding Protein (ABP) domain derived from Streptococcal Protein G.

All SILuPrESTs contain an N-terminal His6ABP fusion tag, used for accurate determination of concentration. The fusion tag sequence is:

GSSHHHHHHSSGLVPRGSHMASLAEAKVLANRELDKYGVSDYHKNLINNAKTVEGVKDLQAQVVESAKKARISEATDGLSDFLKSQTPAEDTVKSIELAEAKVLANRELDKYGVSDYYKNLINNAKTVEGVKALIDEILAALPGTFAHYMDPNSSSVDKLAAA.

The specific human protein sequence begins directly after ′VDKLAAA′.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?