EMAIL THIS PAGE TO A FRIEND
R0529 Sigma-Aldrich

Ro 90-7501

≥98% (HPLC)

Synonym: 2′-(4-Aminophenyl)-[2,5′-bi-1H-benzimidazol]-5-amine

Purchase

Properties

Related Categories Approved Therapeutics/Drug Candidates, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, R,
assay   ≥98% (HPLC)
form   solid
color   brown
solubility   DMSO: ~20 mg/mL
  H2O: insoluble
originator   Roche
SMILES string   Nc1ccc(cc1)-c2nc3ccc(cc3[nH]2)-c4nc5ccc(N)cc5[nH]4
InChI   1S/C20H16N6/c21-13-4-1-11(2-5-13)19-23-15-7-3-12(9-17(15)25-19)20-24-16-8-6-14(22)10-18(16)26-20/h1-10H,21-22H2,(H,23,25)(H,24,26)
InChI key   PAGZCEHLFCJSPV-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Inhibitor of Aß42 fibril formation

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?