• USA Home
  • R2380 - Ribonucleic acid, transfer from Human Placenta

EMAIL THIS PAGE TO A FRIEND
R2380 Sigma

Ribonucleic acid, transfer from Human Placenta

lyophilized powder

Purchase

Properties

form   lyophilized powder
storage temp.   −20°C

Description

Analysis Note

Each product is evaluated for its amino acid acceptor activity using aminoacyl-tRNA synthetase and a mixture of amino acids Ala, Asp, Gly, Ile, Lys, Pro, Thr, Val, Arg, Glu, His, Leu, Phe, Ser, Tyr.

Unit Definition

One unit will yield an A260 of 1.0 in 1.0 mL of water (1 cm light path).

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?