EMAIL THIS PAGE TO A FRIEND
R2502 Sigma-Aldrich

Rubidium iodide

≥99.8% (titration)

Purchase

Properties

assay   ≥99.8% (titration)
density   3.55 g/mL at 25 °C (lit.)
SMILES string   [Rb+].[I-]
InChI   1S/HI.Rb/h1H;/q;+1/p-1
InChI key   WFUBYPSJBBQSOU-UHFFFAOYSA-M
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
RTECS 
VL8925000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

251445 Rubidium iodide, 99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?