• USA Home
  • R5027 - Anti-RIP140 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
R5027 Sigma-Aldrich

Anti-RIP140 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-Receptor-interacting Protein 140

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies against Proteins/Bioactives/Markers/Receptors for Stem Cell Biology, Antibodies for Cell Biology, Antibodies for Epigenetics,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 2 μg/mL
species reactivity   human
mol wt   antigen 140 kDa
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
UniProt accession no.   P48552
Gene Information   human ... NRIP1(8204)

Description

Immunogen

synthetic peptide corresponding to amino acid residues 718-735 of human RIP140.

Target description

RIP140, a nuclear coactivator protein, enhances estrogen receptor transcriptional activity through the AF2 activationdomain in an agonist dependent manner. RIP140 is involved in other steroid receptor pathways, including the retinoid pathway.

Physical form

Solution in phosphate buffered saline containing 1 mg/ml bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Retinoic Acid and Gene Expression

All-trans retinoic acid (RA, ATRA) is a pleiotropic activation factor that regulates genes associated with normal vertebrate cellular processes such as cell differentiation, cell proliferation, apopt...
BioFiles 2008, 3.9, 12.
Keywords: Apoptosis, Cancer, Cell proliferation, Cellular processes, Chromatin immunoprecipitation, Condensations, Degradations, Diseases, Gene expression, Hormones, Immunoprecipitation, Ligands, Metabolism, Oxidations, Phosphorylations, Polymerase chain reaction, Sequences, Teratogens, Transcription, Type, Ubiquitinations, Usage, Vitamins

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?