• USA Home
  • R6251 - Potassium rhodizonate

EMAIL THIS PAGE TO A FRIEND
R6251 Sigma-Aldrich

Potassium rhodizonate

powder

Synonym: 3,4,5,6-Tetraoxocyclohexene-1,2-diol dipotassium salt, Rhodizonic acid potassium salt

Purchase

Properties

InChI Key   NEWIILAQDSMNML-UHFFFAOYSA-L
form   powder
color   dark brown to black
mp   >300 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

392340 Potassium rhodizonate, technical grade, 85%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?