• USA Home
  • R8013 - Stu I from Streptomyces tubercidicus

EMAIL THIS PAGE TO A FRIEND
R8013 Sigma

Stu I from Streptomyces tubercidicus

buffered aqueous glycerol solution

Purchase

Properties

form   buffered aqueous glycerol solution
concentration   10,000 units/mL
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Supplied with 10x Restriction Endonuclease Buffer SB (B8781).

Comment: Overlapping dcm methylation will block cleavage by Stu I.

Specificity

Recognition sequence: 5′-AGG/CCT-3′
Ligation and recutting results: After 2-10-fold Stu I overdigestion of 1 μg λ DNA substrate, results in 100% cutting, >90% of fragments can be ligated, and >95% recut.
Heat inactivation: Inactivated at 65 °C for 20 minutes.

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 0.1 mM EDTA, 100 mM NaCl, 10 mM 2-mercaptoethanol, 50% glycerol (v/v), 0.01% Triton X-100 (v/v) at 4 °C

Safety & Documentation

Safety Information

WGK Germany 
1
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

incubation buffer

Product #

Description

Add to Cart

B8781 Restriction Enzyme Buffer SB, 10 ×

related product

Product #

Description

Add to Cart

LIG2 QuickLink DNA Ligation Kit, joins blunt-end and sticky-end DNA fragments
D2886 DNA Ligase from T4-infected Escherichia coli, buffered aqueous glycerol solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?