EMAIL THIS PAGE TO A FRIEND
R8151 Sigma-Aldrich

RIM-1

≥90% (HPLC)

  • Empirical Formula (Hill Notation) C51H50N6O8S2

  • Molecular Weight 939.11

  •  MDL number MFCD00236454

  •  PubChem Substance ID 329824035

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Q-R, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   DOSMHBDKKKMIEF-UHFFFAOYSA-N
assay   ≥90% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Fluorescent indicator for protein kinase C.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?