• USA Home
  • R8519 - Monoclonal Anti-human CD57/hnk-1

EMAIL THIS PAGE TO A FRIEND
R8519 Sigma

Monoclonal Anti-human CD57/hnk-1

clone vc1.1, whole antiserum

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, HU-HZ, Primary Antibodies
antibody form   whole antiserum
clone   vc1.1, monoclonal
conjugate   unconjugated
shipped in   wet ice
storage temp.   2-8°C

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?