EMAIL THIS PAGE TO A FRIEND
R9151 Sigma

Rimantadine

solid

Synonym: α-Methyl-1-adamantanemethylamine, remantadine

  • CAS Number 13392-28-4

  • Empirical Formula (Hill Notation) C12H21N

  • Molecular Weight 179.30

Purchase

Properties

form   solid
mol wt   apparent mol wt 179.3

Description

Application

Antiviral.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
AU4716000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?