• USA Home
  • R9775 - Anti-RAIDD, Internal Domain antibody produced in rabbit

R9775 Sigma

Anti-RAIDD, Internal Domain antibody produced in rabbit

~0.5 mg/mL, IgG fraction of antiserum

Synonym: Anti-CRADD, Anti-RIP-associated ICH-1/CED-3-homologous protein with DD



Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Adaptors,
species reactivity   human
application(s)   microarray: suitable
  western blot: 0.5-1 μg/mL using HeLa or K562 cell lysates
clone   polyclonal
concentration   ~0.5 mg/mL
antibody form   IgG fraction of antiserum
mol wt   antigen mol wt 22 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CRADD(8738)
biological source   rabbit
conjugate   unconjugated



synthetic peptide corresponding to amino acids 99-117 of human RAIDD.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Price and Availability

Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  -
Protocols & Articles


Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy


Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?