• USA Home
  • RAB1236 - Feline NCAM-1 ELISA

EMAIL THIS PAGE TO A FRIEND
RAB1236 Sigma-Aldrich

Feline NCAM-1 ELISA

for serum, plasma and cell culture supernatants

  •  NACRES NA.32

Purchase

Properties

Related Categories Antibodies, Antibody Kits, ELISA Kits, Sandwich ELISAs
species reactivity   feline
application(s)   ELISA: suitable
input   sample type plasma
sample type cell culture supernatant(s)
sample type serum
assay range   inter-assay cv: <12%
intra-assay cv: <10%
shipped in   wet ice
storage temp.   −20°C
Gene Information   feline ... NCAM1(493956)

Description

General description

This ELISA antibody pair detects Feline Neural Cell Adhesion Molecule 1 (CD56)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?