• USA Home
  • RES0926S-A7 - Sodium chloride

EMAIL THIS PAGE TO A FRIEND
RES0926S-A7 SAFC

Sodium chloride

PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.

Synonym: Halite, Sodium chloride

Purchase

Properties

Related Categories Essential Chemicals, Research Essentials, Salts, Sodium Chloride (NaCl) and Sodium Chloride Solutions, Sodium Salts More...
Quality Level   GMP-COMPENDIA
biological source   synthetic
grade   USP
vapor pressure   1.33 at 865 °C hPa
InChI Key   FAPWRFPIFSIZLT-UHFFFAOYSA-M
product line   PharmaGrade
assay   99.0-100.5% dry basis
form   powder
impurities   Heavy metals; tested, Trace Cations; tested, Trace Anions: tested
loss   ≤0.5% loss on drying
pH   7
mp   801 °C (lit.)
solubility   water: soluble (358 g/l at 20 °C (68 °F))
suitability   suitable for manufacturing use

Description

Linkage

To request the Enhanced Quality Product Documentation for this product, submit a PharmaGrade Product Information Request Form. Please note that access to the Enhanced Quality Product Documentation for this product requires a confidentiality disclosure agreement.

Packaging

Product is available in the following package sizes:
RES0926S-A702X: 1 kg container
RES0926S-A704X: 10 kg container
RES0926S-A705X: 25 kg container

General description

PharmaGrade products are manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production. For more information on the PharmaGrade Program, visit sigma-aldrich.com/safc-pharmagrade.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
VZ4725000
Protocols & Articles

Related Content

Enhanced Quality Program [VIDEO]

The Enhanced Quality Program is a quality management system that is used to differentiate products based on the required level of transparency, control and documentation support determined by the nee...

One Quality Policy [VIDEO]

Sigma-Aldrich maintains one quality policy on a global level to provide consistency in vision, maintained across all manufacturing and distribution sites so that the company on the whole is adhering ...

Quality Management Systems [VIDEO]

See how we combine the use of various Quality Management Systems to manage manufacturing and supply chain for our products, as well as ensure data integrity for information provided to customers via ...

Risk Management [VIDEO]

Our Quality Policy means that we are able to provide certain information to end customers based on a product’s Enhanced Quality level, and that information is limited to what we are able to guarantee...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?