EMAIL THIS PAGE TO A FRIEND
RES1151B-B7 SAFC

Bicine

PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture

Synonym: N,N-Bis(2-hydroxyethyl)glycine, BICINE

  • CAS Number 150-25-4

  • Empirical Formula (Hill Notation) C6H13NO4

  • Molecular Weight 163.17

  •  Beilstein/REAXYS Number 1769362

  •  EC Number 205-755-1

  •  MDL number MFCD00004295

  •  PubChem Substance ID 329824252

  •  NACRES NA.21

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Good’s Buffers, PharmaGrade Buffers
Quality Level   400
biological source   synthetic
Agency/Method   non-compendial
product line   PharmaGrade
form   powder
application(s)   cell culture | mammalian: suitable
impurities   Endotoxin and microbial; tested
useful pH range   7.6 - 9.0
pKa (25 °C)   8.3
suitability   suitable for manufacturing use
foreign activity   Cytotoxicity, DNase, NICKase, RNase, and Protease; tested
SMILES string   OCCN(CCO)CC(O)=O
InChI   1S/C6H13NO4/c8-3-1-7(2-4-9)5-6(10)11/h8-9H,1-5H2,(H,10,11)
InChI key   FSVCELGFZIQNCK-UHFFFAOYSA-N

Description

General description

PharmaGrade products are manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production. For more information on the PharmaGrade Program, visit sigma-aldrich.com/safc-pharmagrade.

Application

Bicine is a biological buffer often referred to as a “Good’s” buffer. The pKa of Bicine is 8.3 which makes Bicine is a good candidate for biological applications that require pH control above physiological. Bicine buffers are commonly used in electrophoresis and protein separations. Bicine is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.

Bicine can be used in a wide array of biological applications including as a component in media formulations. Specific applications include buffers for electrophoresis, protein purification and diagnostic reagent production.

Recommended buffer for low temperature biochemical work. Used in the preparation of stable substrate solution for serum guanase determination. Spacer in plasma protein fractionation with Sephadex® by isotachophoresis.

Packaging

Product is available in the following package sizes:
RES1151B-B701X: 100 gm container
RES1151B-B702X: 1 kg container
RES1151B-B704X: 10 kg container

Linkage

Enhanced Quality Product Documentation on PharmaGrade Bicine

Legal Information

Sephadex is a registered trademark of GE Healthcare

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

pHast Pack
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Enhanced Quality Program [VIDEO]

The Enhanced Quality Program is a quality management system that is used to differentiate products based on the required level of transparency, control and documentation support determined by the nee...

One Quality Policy [VIDEO]

Sigma-Aldrich maintains one quality policy on a global level to provide consistency in vision, maintained across all manufacturing and distribution sites so that the company on the whole is adhering ...

Quality Management Systems [VIDEO]

See how we combine the use of various Quality Management Systems to manage manufacturing and supply chain for our products, as well as ensure data integrity for information provided to customers via ...

Risk Management [VIDEO]

Our Quality Policy means that we are able to provide certain information to end customers based on a product’s Enhanced Quality level, and that information is limited to what we are able to guarantee...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?