• USA Home
  • S0778 - Sodium sulfate decahydrate

EMAIL THIS PAGE TO A FRIEND
S0778 Sigma-Aldrich

Sodium sulfate decahydrate

Synonym: Glauber’s salt

Purchase

Properties

density   1.46 g/mL at 25 °C (lit.)
storage temp.   2-8°C
SMILES string   O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O
InChI   1S/2Na.H2O4S.10H2O/c;;1-5(2,3)4;;;;;;;;;;/h;;(H2,1,2,3,4);10*1H2/q2*+1;;;;;;;;;;;/p-2
InChI key   RSIJVJUOQBWMIM-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?