EMAIL THIS PAGE TO A FRIEND
S1253 Sigma-Aldrich

1-Bromooctadecane

~98% (capillary GC)

Synonym: Octadecyl bromide, Stearyl bromide

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
assay   ~98% (capillary GC)
bp   214-216 °C/12 mmHg (lit.)
mp   25-30 °C (lit.)
density   0.976 g/mL at 25 °C (lit.)
storage temp.   2-8°C
SMILES string   CCCCCCCCCCCCCCCCCCBr
InChI   1S/C18H37Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-18H2,1H3
InChI key   WSULSMOGMLRGKU-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?