• USA Home
  • S1754 - Sodium sesquicarbonate

EMAIL THIS PAGE TO A FRIEND
S1754 Sigma-Aldrich

Sodium sesquicarbonate

  • CAS Number 6106-20-3

  • Linear Formula Na2CO3 · NaHCO3 · 2H2O

  • Molecular Weight 226.03

  •  MDL number MFCD00036143

Purchase

Properties

SMILES string   O.[Na].OC(O)=O
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
FG2099000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?