EMAIL THIS PAGE TO A FRIEND
S2632 Sigma

Sera from rabbit

lyophilized powder (from clotted whole blood)

Purchase

Properties

form   lyophilized powder (from clotted whole blood)
storage temp.   2-8°C

Description

Application

Rabbit serum is used to support cultures of Borrelia burgdorferi, the causative agent of Lyme disease.

Not tested for use in cell culture, see R450

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?