EMAIL THIS PAGE TO A FRIEND
S3170 Sigma-Aldrich

Spin column-200

Purchase

Description

Application

For applications where purified sample is to be stored in low-salt buffer.

Resin-packed microcentrifuge column used to purify DNA >400 bp with 80% recovery, down to 150 bp with reduced recovery; also used to remove PCR primers <30 bases, enzymes, and proteins <1000 kDa.

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?