• USA Home
  • S5000 - DL-Serine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
S5000 Sigma-Aldrich

DL-Serine methyl ester hydrochloride

Purchase

Properties

Related Categories Amino Acids, Amino Acids & Derivatives, Biochemicals and Reagents, Chemical Biology, Chemical Synthesis,
InChI Key   NDBQJIBNNUJNHA-UHFFFAOYSA-N
mp   134-136 °C (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?